Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772003.1 | XP_008770225.1 | 5 - 6 | 756 | 103 | |
XM_214609.8 | XP_214609.6 | 5 - 6 | 756 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |