Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034995.1 | NP_001030167.1 | 8 - 9 | 1621 | 132 | |
XM_006254636.2 | XP_006254698.1 | 8 - 9 | 1715 | 132 | ||
XM_006254637.2 | XP_006254699.1 | 8 - 9 | 2029 | 132 | ||
XM_006254639.2 | XP_006254701.1 | 9 - 10 | 1826 | 132 | ||
XM_006254640.2 | XP_006254702.1 | 7 - 8 | 1565 | 132 | ||
XM_006254641.2 | XP_006254703.1 | 6 - 7 | 1231 | 132 | ||
XM_008772069.1 | XP_008770291.1 | 8 - 9 | 1581 | 132 | ||
GenBank mRNA | BC103648.1 | - | 8 - 9 | 1621 | 7315158 | 132 |
BC166497.1 | - | 5 - 6 | 1092 | 132 | ||
FQ212790.1 | - | 2 - 3 | 215 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |