Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254636.2 | XP_006254698.1 | 1 - 2 | 155 | 77 | |
XM_006254639.2 | XP_006254701.1 | 1 - 2 | 157 | 77 | ||
XM_006254640.2 | XP_006254702.1 | 1 - 2 | 157 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |