Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024373.1 | NP_077349.1 | 2 - 3 | 408 | 84 | |
XM_006254471.2 | XP_006254533.1 | 3 - 4 | 1829 | 84 | ||
GenBank mRNA | U35890.1 | - | 2 - 3 | 408 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |