Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012122.1 | NP_001012122.1 | 3 - 4 | 453 | 77 | |
XM_006242209.2 | XP_006242271.1 | 3 - 4 | 720 | 77 | ||
XM_008765730.1 | XP_008763952.1 | 3 - 4 | 720 | 77 | ||
XM_008765731.1 | XP_008763953.1 | 3 - 4 | 720 | 77 | ||
XM_008765732.1 | XP_008763954.1 | 3 - 4 | 720 | 77 | ||
XM_008765733.1 | XP_008763955.1 | 3 - 4 | 672 | 77 | ||
GenBank mRNA | BC089830.1 | - | 3 - 4 | 453 | 7323197 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |