Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145771.2 | NP_665714.2 | 9 - 10 | 750 | 75 | |
XM_008765674.1 | XP_008763896.1 | 9 - 10 | 934 | 75 | ||
GenBank mRNA | BC078840.1 | - | 9 - 10 | 750 | 7110224 | 75 |
FQ219187.1 | - | 8 - 9 | 739 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |