Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085485.1 | NP_001078954.1 | 5 - 6 | 1467 | 70 | |
GenBank mRNA | BC166588.1 | - | 5 - 6 | 1365 | 7929890 | 70 |
DV724467.1 | - | 1 - 2 | 273 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |