Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108992.1 | NP_001102462.1 | 8 - 9 | 1098 | 82 | |
XM_006242230.1 | XP_006242292.1 | 8 - 9 | 1095 | 82 | ||
XM_006242231.1 | XP_006242293.1 | 7 - 8 | 1038 | 82 | ||
XM_006242233.1 | XP_006242295.1 | 7 - 8 | 891 | 82 | ||
XM_006242234.1 | XP_006242296.1 | 6 - 7 | 834 | 82 | ||
XM_006242235.1 | XP_006242297.1 | 7 - 8 | 678 | 82 | ||
XM_006242236.2 | XP_006242298.1 | 8 - 9 | 862 | 82 | ||
GenBank mRNA | AY724510.1 | - | 6 - 7 | 774 | 82 | |
BC166484.1 | - | 6 - 7 | 627 | 82 | ||
FQ211850.1 | - | 2 - 3 | 182 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |