Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012115.1 | NP_001012115.1 | 7 - 8 | 944 | 96 | |
GenBank mRNA | BC089877.1 | - | 7 - 8 | 944 | 7325489 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |