Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241090.1 | XP_006241152.1 | 12 - 13 | 1546 | 76 | |
XM_006241091.2 | XP_006241153.1 | 12 - 13 | 1655 | 76 | ||
XM_006241092.1 | XP_006241154.1 | 11 - 12 | 1458 | 76 | ||
XM_008765183.1 | XP_008763405.1 | 11 - 12 | 1886 | 76 | ||
XM_234837.7 | XP_234837.3 | 12 - 13 | 1987 | 76 | ||
GenBank mRNA | FQ210490.1 | - | 12 - 13 | 1551 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |