Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773103.1 | XP_008771325.1 | 3 - 4 | 716 | 104 | |
XM_008773104.1 | XP_008771326.1 | 3 - 4 | 727 | 104 | ||
XM_008773105.1 | XP_008771327.1 | 4 - 5 | 901 | 104 | ||
XM_008773106.1 | XP_008771328.1 | 4 - 5 | 952 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |