Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014187.1 | NP_001014209.1 | 4 - 5 | 683 | 72 | |
XM_006236580.2 | XP_006236642.2 | 4 - 5 | 545 | 72 | ||
XM_006236581.2 | XP_006236643.1 | 4 - 5 | 692 | 72 | ||
GenBank mRNA | BC079412.1 | - | 4 - 5 | 683 | 7113320 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Rattus norvegicus Probe spans exons |