Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126296.2 | NP_001119768.2 | 1 - 2 | 200 | 79 | |
XM_006236464.2 | XP_006236526.1 | 1 - 2 | 219 | 79 | ||
XM_006236465.2 | XP_006236527.1 | 1 - 2 | 219 | 79 | ||
XM_006236466.2 | XP_006236528.1 | 1 - 2 | 219 | 79 | ||
GenBank mRNA | BC161978.1 | - | 1 - 2 | 200 | 9029930 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |