Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC088347.1 | - | 2 - 3 | 121 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |