Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191860.1 | NP_001178789.1 | 113 | 128 |
Gene Symbol: LOC100910079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235966.2 | XP_006236028.1 | 1 - 2 | 169 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |