Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773413.1 | XP_008771635.1 | 1 - 2 | 252 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |