Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tectb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106200.1 | NP_001099670.1 | 2 - 3 | 393 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |