Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127681.1 | NP_001121153.1 | 2 - 3 | 321 | 114 | |
XM_006231682.1 | XP_006231744.1 | 2 - 3 | 313 | 114 | ||
GenBank mRNA | BC167081.1 | - | 2 - 3 | 290 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |