Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031777.2 | NP_113965.1 | 3 - 4 | 188 | 77 | |
XM_006250266.2 | XP_006250328.1 | 3 - 4 | 373 | 77 | ||
XM_008769765.1 | XP_008767987.1 | 1 - 2 | 55 | 77 | ||
GenBank mRNA | BC088849.1 | - | 3 - 4 | 188 | 7313699 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |