Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Disp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105983.1 | NP_001099453.1 | 8 - 9 | 1324 | 69 | |
XM_006250381.2 | XP_006250443.1 | 7 - 8 | 1193 | 69 | ||
XM_006250383.2 | XP_006250445.1 | 6 - 7 | 1654 | 69 | ||
XM_006250384.2 | XP_006250446.1 | 6 - 7 | 1133 | 69 | ||
XM_008769827.1 | XP_008768049.1 | 9 - 10 | 2018 | 69 | ||
XM_008769828.1 | XP_008768050.1 | 7 - 8 | 1304 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |