Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053691.2 | NP_446143.1 | 3 - 4 | 682 | 97 | |
GenBank mRNA | AF352021.1 | - | 3 - 4 | 548 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |