Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105987.1 | NP_001099457.1 | 6 - 7 | 524 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |