Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762548.1 | XP_008760770.1 | 1 - 2 | 1039 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |