Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214459.2 | NP_999624.1 | 7 - 8 | 1212 | 65 | |
GenBank mRNA | AY500157.1 | - | 7 - 8 | 1212 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |