Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031803.1 | NP_113991.1 | 5 - 6 | 568 | 69 | |
GenBank mRNA | AF059273.1 | - | 5 - 6 | 568 | 69 | |
AF205778.1 | - | 462 | 69 | |||
AF205779.1 | - | 462 | 69 | |||
AF205780.1 | - | 462 | 69 | |||
FQ211504.1 | - | 612 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |