Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729258.4 | XP_002729304.3 | 8 - 9 | 1364 | 103 | |
XM_006235827.2 | XP_006235889.1 | 9 - 10 | 1481 | 103 | ||
XM_006235828.2 | XP_006235890.1 | 8 - 9 | 1404 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |