Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baiap2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034140.1 | NP_001029312.1 | 5 - 6 | 574 | 174 | |
GenBank mRNA | BC089216.1 | - | 4 - 5 | 417 | 174 | |
BC105815.1 | - | 5 - 6 | 574 | 7367101 | 174 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |