Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105912.1 | NP_001099382.1 | 7 - 8 | 934 | 127 | |
XM_006248920.2 | XP_006248982.1 | 7 - 8 | 917 | 127 | ||
XM_006248921.2 | XP_006248983.1 | 7 - 8 | 1317 | 127 | ||
XM_006248923.2 | XP_006248985.1 | 7 - 8 | 1190 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |