Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105736.2 | NP_001099206.1 | 1 - 2 | 165 | 128 | |
GenBank mRNA | BE109715.1 | - | 2 - 3 | 143 | 128 | |
EU202679.1 | - | 1 - 2 | 111 | 128 | ||
EU202680.1 | - | 1 - 2 | 111 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |