Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034198.2 | NP_001029370.1 | 6 - 7 | 694 | 147 | |
NM_021671.3 | NP_067703.1 | 6 - 7 | 694 | 147 | ||
GenBank mRNA | AB006135.1 | - | 6 - 7 | 652 | 147 | |
CK359233.1 | - | 6 - 7 | 575 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |