Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130822.1 | NP_570835.1 | 23 - 24 | 3914 | 57 | |
GenBank mRNA | AF063103.1 | - | 23 - 24 | 3914 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Rattus norvegicus Probe spans exons |