Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199374.2 | NP_955406.2 | 9 - 10 | 1607 | 95 | |
XM_006250891.2 | XP_006250953.1 | 9 - 10 | 1618 | 95 | ||
GenBank mRNA | AB116526.1 | - | 9 - 10 | 1557 | 95 | |
BC078996.1 | - | 9 - 10 | 1607 | 7112725 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |