Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191596.1 | NP_001178525.1 | 4 - 5 | 635 | 79 | |
XM_006250888.2 | XP_006250950.1 | 4 - 5 | 1145 | 79 | ||
XM_006250889.2 | XP_006250951.1 | 4 - 5 | 1198 | 79 | ||
XM_008770137.1 | XP_008768359.1 | 4 - 5 | 1228 | 79 | ||
GenBank mRNA | FQ211578.1 | - | 4 - 5 | 683 | 79 | |
GU455413.1 | - | 4 - 5 | 559 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |