Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013989.1 | NP_001014011.1 | 7 - 8 | 1106 | 77 | |
GenBank mRNA | BC079225.1 | - | 7 - 8 | 1106 | 7132309 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |