Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100889.1 | NP_001094359.1 | 3 - 4 | 448 | 151 | |
XM_006250723.2 | XP_006250785.1 | 3 - 4 | 1459 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |