Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014233.2 | NP_001014255.1 | 6 - 7 | 811 | 89 | |
XM_006251888.1 | XP_006251950.1 | 6 - 7 | 784 | 89 | ||
XM_006251889.2 | XP_006251951.1 | 5 - 6 | 670 | 89 | ||
GenBank mRNA | BC087604.1 | - | 6 - 7 | 814 | 7312219 | 89 |
FQ225412.1 | - | 6 - 7 | 789 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |