Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100510.1 | NP_001093980.1 | 2 - 3 | 330 | 110 | |
GenBank mRNA | BC087140.1 | - | 2 - 3 | 254 | 110 | |
BC158592.1 | - | 2 - 3 | 330 | 8362956 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |