Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt16h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107261.1 | NP_001100731.1 | 23 - 24 | 2855 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |