Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105981.1 | NP_001099451.1 | 2 - 3 | 229 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |