Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135119.2 | NP_001128591.2 | 3 - 4 | 647 | 75 | |
XM_006250462.2 | XP_006250524.1 | 4 - 5 | 652 | 75 | ||
XM_008769851.1 | XP_008768073.1 | 3 - 4 | 1316 | 75 | ||
XM_008769852.1 | XP_008768074.1 | 3 - 4 | 649 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |