Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109564.1 | NP_001103034.1 | 1 | 3024 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |