Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppdpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235747.2 | XP_006235809.1 | 1 - 2 | 251 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |