Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020976.1 | NP_066125.1 | 3 - 4 | 210 | 73 | |
GenBank mRNA | AF178086.1 | - | 3 - 4 | 210 | 73 | |
BC078838.1 | - | 3 - 4 | 267 | 7129929 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |