Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191732.1 | NP_001178661.1 | 3 - 4 | 520 | 89 | |
XM_006256834.2 | XP_006256896.1 | 3 - 4 | 494 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |