Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004236.1 | NP_001004236.1 | 3 - 4 | 391 | 79 | |
XM_006238639.1 | XP_006238701.1 | 5 - 6 | 664 | 79 | ||
GenBank mRNA | BC078938.1 | - | 3 - 4 | 391 | 7108356 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |