Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177931.2 | NP_808792.1 | 10 - 11 | 1711 | 123 | |
XM_006238515.2 | XP_006238577.1 | 11 - 12 | 1913 | 123 | ||
XM_006238516.2 | XP_006238578.1 | 12 - 13 | 2026 | 123 | ||
XM_006238518.2 | XP_006238580.1 | 11 - 12 | 1913 | 123 | ||
GenBank mRNA | AB105378.1 | - | 10 - 11 | 1711 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |