Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106679.2 | NP_001100149.2 | 1 - 2 | 290 | 107 | |
XM_006238640.2 | XP_006238702.1 | 2 - 3 | 462 | 107 | ||
XM_006238642.2 | XP_006238704.1 | 2 - 3 | 473 | 107 | ||
XM_006238643.2 | XP_006238705.1 | 2 - 3 | 474 | 107 | ||
XM_006238644.2 | XP_006238706.1 | 2 - 3 | 474 | 107 | ||
XM_006238645.2 | XP_006238707.1 | 2 - 3 | 474 | 107 | ||
XM_008763992.1 | XP_008762214.1 | 2 - 3 | 473 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |