Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025123.3 | NP_001020294.3 | 3 - 4 | 692 | 88 | |
GenBank mRNA | BC093594.1 | - | 3 - 4 | 692 | 7314531 | 88 |
BC100617.1 | - | 3 - 4 | 691 | 7934552 | 88 | |
FQ211817.1 | - | 3 - 4 | 593 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |