Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106820.1 | NP_001100290.1 | 15 - 16 | 2383 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |