Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017281.1 | NP_058977.1 | 8 - 9 | 729 | 89 | |
GenBank mRNA | BC063170.1 | - | 8 - 9 | 749 | 6921964 | 89 |
FQ213330.1 | - | 4 - 5 | 362 | 89 | ||
FQ215861.1 | - | 4 - 5 | 362 | 89 | ||
FQ216011.1 | - | 4 - 5 | 362 | 89 | ||
FQ216241.1 | - | 4 - 5 | 362 | 89 | ||
FQ220234.1 | - | 4 - 5 | 361 | 89 | ||
FQ220280.1 | - | 4 - 5 | 362 | 89 | ||
FQ226777.1 | - | 4 - 5 | 362 | 89 | ||
FQ234380.1 | - | 4 - 5 | 362 | 89 | ||
X53304.1 | - | 8 - 9 | 729 | 89 | ||
X55986.1 | - | 8 - 9 | 685 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |